Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpr-B
Entrez Gene ID
41357
Gene Name
SCP-containing protein B
Gene Aliases
CG17210, Dmel\CG17210, Dmel_CG17210, SCPR-B, anon-WO0140519.115, cg17210, scp-B
Chromosome Location
Chr.3R: 11240041 - 11240991 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037888
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141804.2 NP_650061.1 2 - 2 836 108
GenBank mRNA AY060783.1 - 2 - 2 827 108

View Full Product Details