Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sta
- Entrez Gene ID
-
31106
- Gene Name
- stubarista
- Gene Aliases
-
37LRP/p40, CG14792, Dmel\CG14792, Dmel_CG14792, EG:80H7.6, Lam-r, P40, Rp 40, Rp40, RpSA, STA, Sa, Sta, anon-EST:Liang-2.56, chrX:1339078..1339216, clone 2.56, k14, l(1)2Af, l(1)G0130, l(1)G0448
- Chromosome Location
-
Chr.X: 1481797 - 1483413 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003517
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166890.3 |
NP_726745.3 |
1 - 1 |
137 |
91 |
View Full Product Details