Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
adgrv1
Entrez Gene ID
415105
Gene Name
adhesion G protein-coupled receptor V1
Gene Aliases
gpr98, mass1, vlgr1
Chromosome Location
Chr.5: 48683336 - 48940174 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001845.2 NP_001001845.2 72 - 73 15102 122
XM_005165511.4 XP_005165568.1 72 - 73 15221 122
XM_009301753.3 XP_009300028.1 73 - 74 15243 122
GenBank mRNA AY540131.1 - 72 - 73 15106 122

View Full Product Details