Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dtd1
Entrez Gene ID
445046
Gene Name
D-aminoacyl-tRNA deacylase 1
Gene Aliases
hars2, zgc:92657
Chromosome Location
Chr.6: 6802556 - 6808505 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003440.1 NP_001003440.1 3 - 4 400 107
GenBank mRNA BC076141.1 - 3 - 4 400 107

View Full Product Details