Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hoxc9a
Entrez Gene ID
58043
Gene Name
homeobox C9a
Gene Aliases
BZ-39, bz39, hoxc9, zgc:109844
Chromosome Location
Chr.23: 36095306 - 36097769 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131528.3 NP_571603.3 1 - 2 595 67
GenBank mRNA BC095014.1 - 1 - 2 566 67
BC164046.1 - 1 - 2 566 67
DQ060556.1 - 1 - 2 244 67
EH591454.1 - 1 - 2 594 67

View Full Product Details