Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
hyi
Entrez Gene ID
393305
Gene Name
hydroxypyruvate isomerase
Gene Aliases
zgc:63850
Chromosome Location
Chr.6: 2619890 - 2627488 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200334.1 NP_956628.1 5 - 6 671 149
GenBank mRNA BC053111.1 - 5 - 6 671 149

View Full Product Details