Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp2k8
Entrez Gene ID
561462
Gene Name
cytochrome P450, family 2, subfamily K, polypeptide 8
Gene Aliases
zgc:194733
Chromosome Location
Chr.3: 12784460 - 12802380 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130623.2 NP_001124095.1 3 - 4 618 71
GenBank mRNA AY772712.1 - 3 - 4 534 71
BC163074.1 - 3 - 4 618 71

View Full Product Details