Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mtmr7b
Entrez Gene ID
559198
Gene Name
myotubularin related protein 7b
Gene Aliases
zgc:114101
Chromosome Location
Chr.1: 129632 - 153856 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030126.1 NP_001025297.1 2 - 3 360 114
XM_005168352.4 XP_005168409.1 2 - 3 388 114
GenBank mRNA BC097161.1 - 2 - 3 360 114

View Full Product Details