Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- dhrs9
- Entrez Gene ID
-
322529
- Gene Name
- dehydrogenase/reductase (SDR family) member 9
- Gene Aliases
-
RDHA, rdh1l, wu:fb64b07, zgc:73286
- Chromosome Location
-
Chr.9: 48706039 - 48736182 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199609.1 |
NP_955903.1 |
2 - 3 |
399 |
106 |
GenBank mRNA |
BC059614.1 |
- |
2 - 3 |
399 |
106 |
- Gene Symbol
- znf622
- Entrez Gene ID
-
407703
- Gene Name
- zinc finger protein 622
- Gene Aliases
-
Zfp622, znf622l
- Chromosome Location
-
Chr.2: 41563073 - 41571494 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AI497329.1 |
- |
1 - 2 |
320 |
106 |
View Full Product Details