Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpp30
Entrez Gene ID
541352
Gene Name
ribonuclease P/MRP 30 subunit
Gene Aliases
fk66h08, wu:fk66h08, zgc:110189
Chromosome Location
Chr.12: 16446227 - 16452475 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013479.1 NP_001013497.1 3 - 4 286 124
XM_021480064.1 XP_021335739.1 3 - 4 139 124
GenBank mRNA BC091449.1 - 3 - 4 286 124
BC164838.1 - 3 - 4 236 124

View Full Product Details