Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttc5
Entrez Gene ID
550347
Gene Name
tetratricopeptide repeat domain 5
Gene Aliases
wu:fi33h05, wu:fy81e07, zgc:112059
Chromosome Location
Chr.7: 250523 - 260613 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017654.1 NP_001017654.1 5 - 6 668 63
GenBank mRNA BC093176.1 - 5 - 6 668 63

View Full Product Details