Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
jade1
Entrez Gene ID
327437
Gene Name
jade family PHD finger 1
Gene Aliases
fi11a04, phf17, wu:fi11a04, zgc:55323
Chromosome Location
Chr.17: 64695 - 83990 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199805.1 NP_956099.1 8 - 9 1080 65
XM_005158491.4 XP_005158548.1 8 - 9 1355 65
XM_005158492.4 XP_005158549.1 8 - 9 1227 65
XM_017351598.2 XP_017207087.1 8 - 9 1599 65
GenBank mRNA BC046874.1 - 8 - 9 1080 65
BC165673.1 - 7 - 8 997 65

View Full Product Details