Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
uqcrc2a
Entrez Gene ID
436930
Gene Name
ubiquinol-cytochrome c reductase core protein 2a
Gene Aliases
fb33d01, fc43c05, wu:fb33d01, wu:fc43c05, zgc:92453
Chromosome Location
Chr.3: 58167269 - 58185030 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002657.2 NP_001002657.2 1 - 2 112 79
GenBank mRNA BC076480.1 - 1 - 2 78 79
BC164681.1 - 1 - 2 69 79
EB969638.1 - 1 - 2 113 79
EH436572.1 - 1 - 2 89 79

View Full Product Details