Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
car15
Entrez Gene ID
568143
Gene Name
carbonic anhydrase 15
Gene Aliases
si:ch211-265m20.6
Chromosome Location
Chr.10: 175673 - 187935 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045139.1 NP_001038604.1 8 - 9 888 153

View Full Product Details