Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mmab
Entrez Gene ID
557077
Gene Name
metabolism of cobalamin associated B
Gene Aliases
si:dkey-189p24.5
Chromosome Location
Chr.5: 19489581 - 19494048 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082809.1 NP_001076278.1 6 - 7 557 98
GenBank mRNA BC133925.1 - 6 - 7 510 98

View Full Product Details