Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rrp15
Entrez Gene ID
327053
Gene Name
ribosomal RNA processing 15 homolog
Gene Aliases
wu:fa20e10, zgc:56269
Chromosome Location
Chr.16: 4196932 - 4203022 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199762.1 NP_956056.1 2 - 3 351 88
GenBank mRNA BC046037.1 - 2 - 3 351 88

View Full Product Details