Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rmnd1
Entrez Gene ID
553810
Gene Name
required for meiotic nuclear division 1 homolog
Gene Aliases
zgc:113022
Chromosome Location
Chr.17: 7795561 - 7819002 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020772.1 NP_001018608.1 11 - 12 1299 74
XM_009292818.3 XP_009291093.1 11 - 12 1539 74
XM_009292819.3 XP_009291094.1 11 - 12 1398 74
XM_021467167.1 XP_021322842.1 11 - 12 1228 74
GenBank mRNA BC095858.1 - 11 - 12 1299 74

View Full Product Details