Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lft2
Entrez Gene ID
30146
Gene Name
lefty2
Gene Aliases
atv2, cb720, fe38b03, wu:fe38b03
Chromosome Location
Chr.17: 8308182 - 8312891 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130961.1 NP_571036.1 3 - 4 772 75
GenBank mRNA AF132445.1 - 3 - 4 772 75
BC097092.1 - 3 - 4 796 75
KM655581.1 - 3 - 4 763 75

View Full Product Details