Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
lamb1a
Entrez Gene ID
286830
Gene Name
laminin, beta 1a
Gene Aliases
cb95, fc20b05, gup, lamb1, sb:cb95, wu:fc20b05
Chromosome Location
Chr.25: 31546828 - 31629095 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173275.1 NP_775382.1 12 - 13 1552 68
GenBank mRNA AF468049.1 - 12 - 13 1552 68

View Full Product Details