Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tra2b
Entrez Gene ID
394172
Gene Name
transformer 2 beta homolog
Gene Aliases
sfrs10, zgc:56612
Chromosome Location
Chr.9: 12579077 - 12594777 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201197.1 NP_957491.1 6 - 7 877 93
XM_005167473.2 XP_005167530.1 6 - 7 93
XM_005167474.4 XP_005167531.1 7 - 8 93
GenBank mRNA BC049051.1 - 6 - 7 877 93

View Full Product Details