Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stmn4l
Entrez Gene ID
436836
Gene Name
stathmin-like 4, like
Gene Aliases
zgc:92760
Chromosome Location
Chr.17: 12698509 - 12709321 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002563.1 NP_001002563.1 5 - 6 619 78
GenBank mRNA BC076237.1 - 5 - 6 619 78

View Full Product Details