Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atp6v0a1a
Entrez Gene ID
324307
Gene Name
ATPase H+ transporting V0 subunit a1a
Gene Aliases
atp6v0a1, wu:fc25b09, zgc:76965
Chromosome Location
Chr.3: 16922165 - 16973129 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212672.1 NP_997837.1 2 - 3 192 76
XM_005163798.3 XP_005163855.1 2 - 3 304 76
GenBank mRNA BC066692.1 - 2 - 3 192 76

View Full Product Details