Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cpped1
Entrez Gene ID
492772
Gene Name
calcineurin-like phosphoesterase domain containing 1
Gene Aliases
zgc:101576
Chromosome Location
Chr.12: 368043 - 432949 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007413.1 NP_001007414.1 1 - 2 213 59
GenBank mRNA BC085372.1 - 1 - 2 213 59

View Full Product Details