Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stim1a
Entrez Gene ID
556360
Gene Name
stromal interaction molecule 1a
Gene Aliases
si:dkey-24p1.5
Chromosome Location
Chr.15: 38221146 - 38297737 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044799.1 NP_001038264.1 2 - 3 397 79
XM_005173392.4 XP_005173449.1 2 - 3 447 79
XM_005173393.4 XP_005173450.1 2 - 3 447 79
XM_009291871.3 XP_009290146.1 2 - 3 447 79
GenBank mRNA BC134856.1 - 2 - 3 359 79

View Full Product Details