Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sptan1
Entrez Gene ID
553383
Gene Name
spectrin alpha, non-erythrocytic 1
Gene Aliases
im:7157190, spna2, wu:fa20e05, wu:fb33g08, wu:fk32a10, zgc:112229
Chromosome Location
Chr.21: 13150896 - 13217905 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098488.1 NP_001091958.1 31 - 32 4239 60
XM_021468823.1 XP_021324498.1 30 - 31 4159 60
XM_021468824.1 XP_021324499.1 31 - 32 4291 60
XM_021468825.1 XP_021324500.1 31 - 32 4370 60
XM_021468826.1 XP_021324501.1 31 - 32 4177 60
XM_021468827.1 XP_021324502.1 29 - 30 4126 60
XM_021468828.1 XP_021324503.1 29 - 30 4111 60
XM_021468829.1 XP_021324504.1 29 - 30 4099 60
XM_021468830.1 XP_021324505.1 30 - 31 4159 60
XM_021468831.1 XP_021324506.1 30 - 31 4159 60
XM_021468832.1 XP_021324507.1 28 - 29 4078 60
XM_021468833.1 XP_021324508.1 29 - 30 4099 60
XM_021468834.1 XP_021324509.1 29 - 30 4099 60
GenBank mRNA EF375552.1 - 31 - 32 4239 60

View Full Product Details