Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sptan1
Entrez Gene ID
553383
Gene Name
spectrin alpha, non-erythrocytic 1
Gene Aliases
im:7157190, spna2, wu:fa20e05, wu:fb33g08, wu:fk32a10, zgc:112229
Chromosome Location
Chr.21: 13150896 - 13217905 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098488.1 NP_001091958.1 33 - 34 4581 118
XM_021468823.1 XP_021324498.1 32 - 33 4501 118
XM_021468824.1 XP_021324499.1 33 - 34 4633 118
XM_021468825.1 XP_021324500.1 33 - 34 4712 118
XM_021468826.1 XP_021324501.1 33 - 34 4519 118
XM_021468827.1 XP_021324502.1 31 - 32 4468 118
XM_021468828.1 XP_021324503.1 31 - 32 4453 118
XM_021468829.1 XP_021324504.1 31 - 32 4441 118
XM_021468830.1 XP_021324505.1 32 - 33 4501 118
XM_021468831.1 XP_021324506.1 32 - 33 4501 118
XM_021468832.1 XP_021324507.1 30 - 31 4420 118
XM_021468833.1 XP_021324508.1 31 - 32 4441 118
XM_021468834.1 XP_021324509.1 31 - 32 4441 118
GenBank mRNA EF375552.1 - 33 - 34 4581 118

View Full Product Details