Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fn1b
Entrez Gene ID
334613
Gene Name
fibronectin 1b
Gene Aliases
cb1057, fn1l, fn3, wu:fa14f11, wu:fb03c02
Chromosome Location
Chr.1: 5504131 - 5543086 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013261.1 NP_001013279.1 8 - 9 1312 75
XM_005160284.3 XP_005160341.1 8 - 9 75
XM_009305554.3 XP_009303829.1 8 - 9 75
GenBank mRNA AY538257.1 - 8 - 9 1312 75
AY725818.1 - 8 - 9 75

View Full Product Details