Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myh11a
Entrez Gene ID
554168
Gene Name
myosin, heavy chain 11a, smooth muscle
Gene Aliases
myh11
Chromosome Location
Chr.6: 8499855 - 8580885 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024448.1 NP_001019619.1 15 - 16 1972 92
XM_021476875.1 XP_021332550.1 16 - 17 2025 92
XM_021476876.1 XP_021332551.1 16 - 17 2025 92
GenBank mRNA BC162152.1 - 15 - 16 1936 92
DQ022376.1 - 15 - 16 1972 92

View Full Product Details