Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
myh11a
Entrez Gene ID
554168
Gene Name
myosin, heavy chain 11a, smooth muscle
Gene Aliases
myh11
Chromosome Location
Chr.6: 8499855 - 8580885 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024448.1 NP_001019619.1 31 - 32 4467 90
XM_021476875.1 XP_021332550.1 32 - 33 4520 90
XM_021476876.1 XP_021332551.1 32 - 33 4520 90
GenBank mRNA BC162152.1 - 31 - 32 4431 90
DQ022376.1 - 31 - 32 4467 90

View Full Product Details