Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
saa
Entrez Gene ID
449557
Gene Name
serum amyloid A
Gene Aliases
zgc:103580
Chromosome Location
Chr.7: 12950507 - 12956434 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005599.2 NP_001005599.1 3 - 4 257 101
GenBank mRNA BC081487.1 - 3 - 4 262 101
BC164922.1 - 2 - 3 260 101

View Full Product Details