Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC103909796
Entrez Gene ID
103909796
Gene Name
golgin subfamily A member 6-like protein 2
Gene Aliases
-
Chromosome Location
Chr.25: 272909 - 278081 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_009297684.3 XP_009295959.1 2 - 2 2506 128
XM_009297685.3 XP_009295960.1 3 - 3 1043 128

View Full Product Details