Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
txndc17
Entrez Gene ID
445062
Gene Name
thioredoxin domain containing 17
Gene Aliases
txnl5, zgc:91920
Chromosome Location
Chr.15: 16328450 - 16332990 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003456.1 NP_001003456.1 2 - 3 326 123
GenBank mRNA BC078373.1 - 2 - 3 326 123
BC164925.1 - 2 - 3 255 123

View Full Product Details