Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
morc3b
Entrez Gene ID
445185
Gene Name
MORC family CW-type zinc finger 3b
Gene Aliases
cb561, sb:cb561, sb:cb69, zgc:101052
Chromosome Location
Chr.1: 47178529 - 47187868 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003579.1 NP_001003579.1 4 - 5 613 64
GenBank mRNA BC078200.1 - 4 - 5 613 64

View Full Product Details