Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnajc27
Entrez Gene ID
402784
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 27
Gene Aliases
rbj, zgc:194263, zgc:194279
Chromosome Location
Chr.17: 33296606 - 33306682 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007162.2 NP_001007163.1 5 - 6 586 114
XM_005158728.4 XP_005158785.1 5 - 6 831 114
GenBank mRNA BC162857.1 - 5 - 6 586 114
BC162873.1 - 5 - 6 586 114

View Full Product Details