Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mul1b
Entrez Gene ID
100004181
Gene Name
mitochondrial E3 ubiquitin protein ligase 1b
Gene Aliases
im:7146383, zgc:165594
Chromosome Location
Chr.23: 37286509 - 37293429 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098760.1 NP_001092230.1 4 - 5 373 69
XM_005166206.4 XP_005166263.1 4 - 5 366 69
GenBank mRNA BC142887.1 - 4 - 5 373 69

View Full Product Details