Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zic2b
Entrez Gene ID
570746
Gene Name
zic family member 2 (odd-paired homolog, Drosophila) b
Gene Aliases
hm:zeh0655, id:ibd5017, zic2.2, zic2l
Chromosome Location
Chr.1: 29858045 - 29860849 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001820.1 NP_001001820.1 1 - 2 1192 77
GenBank mRNA AF207751.2 - 1 - 2 1192 77
BC098556.1 - 1 - 2 1205 77
BC171625.1 - 1 - 2 1188 77
BC171629.1 - 1 - 2 1188 77

View Full Product Details