Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
slco1d1
Entrez Gene ID
326845
Gene Name
solute carrier organic anion transporter family, member 1D1
Gene Aliases
wu:fe36b05, zgc:163027
Chromosome Location
Chr.4: 77389667 - 77432607 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348086.1 NP_001335015.1 7 - 8 966 76
XM_021474933.1 XP_021330608.1 8 - 9 984 76
GenBank mRNA BC139695.1 - 7 - 8 993 76
KP294314.1 - 7 - 8 957 76

View Full Product Details