Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ube2g1b
Entrez Gene ID
100000479
Gene Name
ubiquitin-conjugating enzyme E2G 1b (UBC7 homolog, yeast)
Gene Aliases
ube2g1, wu:fc68b05, wu:fc75g08, wu:fk32c04, zgc:55321
Chromosome Location
Chr.21: 17031635 - 17037902 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213530.1 NP_998695.1 4 - 5 576 59
GenBank mRNA BC045309.1 - 4 - 5 576 59
BC071506.1 - 4 - 5 548 59
BC165416.1 - 4 - 5 459 59
BC171345.1 - 4 - 5 532 59

View Full Product Details