Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPPA
Entrez Gene ID
100034201
Gene Name
natriuretic peptide A
Gene Aliases
CDD
Chromosome Location
Chr.2: 40021300 - 40022688 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082501.2 NP_001075970.1 1 - 2 125 62
GenBank mRNA X58563.1 - - 125 62

View Full Product Details