Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP6R3
Entrez Gene ID
100053490
Gene Name
protein phosphatase 6 regulatory subunit 3
Gene Aliases
SAPS3
Chromosome Location
Chr.12: 31467205 - 31607697 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113235.1 NP_001106706.1 5 - 6 798 116
XM_023653807.1 XP_023509575.1 - 830 116
XM_023653808.1 XP_023509576.1 - 654 116
XM_023653810.1 XP_023509578.1 - 662 116
XM_023653811.1 XP_023509579.1 - 830 116
XM_023653812.1 XP_023509580.1 - 830 116
XM_023653813.1 XP_023509581.1 - 830 116
XM_023653814.1 XP_023509582.1 - 830 116
XM_023653815.1 XP_023509583.1 - 830 116
XM_023653816.1 XP_023509584.1 - 830 116
XM_023653817.1 XP_023509585.1 - 830 116
XM_023653818.1 XP_023509586.1 - 830 116
XM_023653819.1 XP_023509587.1 - 830 116
XM_023653821.1 XP_023509589.1 - 830 116
XM_023653822.1 XP_023509590.1 - 351 116
XM_023653823.1 XP_023509591.1 - 410 116
XM_023653824.1 XP_023509592.1 - 830 116
GenBank mRNA AB371895.1 - 5 - 6 798 116
AB371896.1 - 5 - 6 798 116
AB371897.1 - 5 - 6 798 116

View Full Product Details