Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MATR3
Entrez Gene ID
100062181
Gene Name
matrin 3
Gene Aliases
-
Chromosome Location
Chr.14: 36628273 - 36658757 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205261.1 NP_001192190.1 10 - 11 2070 122
XM_014730313.2 XP_014585799.2 - 2059 122
XM_014730314.2 XP_014585800.2 - 2035 122
XM_014730317.2 XP_014585803.2 - 1909 122
GenBank mRNA CX604359.1 - 3 - 4 319 122

View Full Product Details