Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRIA2
Entrez Gene ID
100069331
Gene Name
glutamate ionotropic receptor AMPA type subunit 2
Gene Aliases
-
Chromosome Location
Chr.2: 76948197 - 77091772 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185087.3 NP_001172016.3 15 - 16 3113 80
NM_001185088.2 NP_001172017.2 15 - 16 3113 80
NM_001185089.2 NP_001172018.2 15 - 16 2629 80

View Full Product Details