Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL18
Entrez Gene ID
421585
Gene Name
mitochondrial ribosomal protein L18
Gene Aliases
-
Chromosome Location
Chr.3: 45688638 - 45691180 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_419623.5 XP_419623.1 3 - 4 603 92
GenBank mRNA BX930290.1 - 3 - 4 575 92
BX930840.1 - 3 - 4 584 92
BX934122.1 - 3 - 4 609 92
BX935429.2 - 3 - 4 588 92
CR406985.1 - 1 - 2 1002 92

View Full Product Details