Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP58
Entrez Gene ID
423882
Gene Name
cilia and flagella associated protein 58
Gene Aliases
C10orf80, CCDC147
Chromosome Location
Chr.6: 25237368 - 25297331 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025151675.1 XP_025007443.1 3 - 4 607 112
XM_421748.4 XP_421748.3 2 - 3 347 112
GenBank mRNA BX933054.2 - 1 - 2 502 112
BX950781.2 - 2 - 3 501 112

View Full Product Details