Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC6
Entrez Gene ID
420112
Gene Name
armadillo repeat containing 6
Gene Aliases
-
Chromosome Location
Chr.28: 3756588 - 3761760 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025144305.1 XP_025000073.1 2 - 3 298 62
XM_418230.6 XP_418230.1 2 - 3 288 62
GenBank mRNA BX931681.1 - 1 - 2 227 62
CR354313.1 - 1 - 2 481 62

View Full Product Details