Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D31
Entrez Gene ID
420349
Gene Name
TBC1 domain family member 31
Gene Aliases
WDR67
Chromosome Location
Chr.2: 137916084 - 137941378 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015283159.2 XP_015138645.1 17 - 18 2515 118
XM_025148148.1 XP_025003916.1 16 - 17 2887 118
XM_025148149.1 XP_025003917.1 16 - 17 2887 118
XM_025148150.1 XP_025003918.1 16 - 17 2399 118
XM_418456.6 XP_418456.2 16 - 17 2490 118
GenBank mRNA CR387563.1 - 2 - 3 282 118

View Full Product Details