Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDCD1
Entrez Gene ID
424913
Gene Name
programmed cell death 1
Gene Aliases
-
Chromosome Location
Chr.9: 15045846 - 15050270 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_422723.6 XP_422723.3 3 - 4 686 69
GenBank mRNA CR390246.1 - 2 - 3 201 69

View Full Product Details