Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCD5
Entrez Gene ID
422598
Gene Name
stearoyl-CoA desaturase 5
Gene Aliases
-
Chromosome Location
Chr.4: 46361217 - 46379016 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015276479.2 XP_015131965.1 3 - 4 478 91
XM_015276480.1 XP_015131966.1 4 - 5 660 91
GenBank mRNA EU275768.1 - 3 - 4 479 91
MG066596.1 - 4 - 5 490 91

View Full Product Details