Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNRF2
Entrez Gene ID
420678
Gene Name
zinc and ring finger 2
Gene Aliases
-
Chromosome Location
Chr.2: 41208589 - 41266458 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015281773.2 XP_015137259.1 2 - 3 1138 136
GenBank mRNA CR353109.1 - 2 - 3 238 136

View Full Product Details