Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YIPF7
Entrez Gene ID
422774
Gene Name
Yip1 domain family member 7
Gene Aliases
-
Chromosome Location
Chr.4: 67636995 - 67650496 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277822.1 NP_001264751.1 2 - 3 150 138
GenBank mRNA BX935861.1 - 2 - 3 151 138

View Full Product Details